Assay Details
| Gene Symbol: AT3G63480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116213.3 | NP_567148.1 | 14 - 15 | 1215 | 125 | |
| NM_180411.3 | NP_850742.1 | 14 - 15 | 1285 | 125 | ||
| GenBank mRNA | AK119094.1 | - | 14 - 15 | 1203 | 125 | |
| BT005391.1 | - | 13 - 14 | 1107 | 125 | ||
More Information
Set Membership: |
|

AT3G63480
Genomic Map