Assay Details
| Gene Symbol: GLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120496.3 | NP_568134.1 | 13 - 14 | 2555 | 71 | |
| NM_180432.2 | NP_850763.1 | 12 - 13 | 2387 | 71 | ||
More Information
Set Membership: |
|

GLU1
Genomic Map