Assay Details
| Gene Symbol: GLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120496.3 | NP_568134.1 | 2 - 3 | 831 | 104 | |
| NM_180432.2 | NP_850763.1 | 1 - 2 | 663 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLU1| Gene Symbol: GLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120496.3 | NP_568134.1 | 2 - 3 | 831 | 104 | |
| NM_180432.2 | NP_850763.1 | 1 - 2 | 663 | 104 | ||
Set Membership: |
|