Assay Details
| Gene Symbol: GLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120496.3 | NP_568134.1 | 25 - 26 | 4079 | 105 | |
| NM_180432.2 | NP_850763.1 | 24 - 25 | 3911 | 105 | ||
More Information
Set Membership: |
|

GLU1
Genomic Map