Assay Details
| Gene Symbol: UBP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125707.2 | NP_001119179.1 | 22 - 23 | 2574 | 92 | |
| NM_120743.3 | NP_568171.1 | 22 - 23 | 2747 | 92 | ||
| NM_180452.1 | NP_850783.1 | 22 - 23 | 2573 | 92 | ||
| GenBank mRNA | AF360198.1 | - | 22 - 23 | 2573 | 92 | |
| AY142616.1 | - | 22 - 23 | 2462 | 92 | ||
More Information
Set Membership: |
|

UBP12
Genomic Map