Assay Details
| Gene Symbol: GRP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120836.5 | NP_196371.1 | 1 - 2 | 299 | 71 | |
| NM_180457.3 | NP_850788.2 | 1 - 2 | 299 | 71 | ||
| GenBank mRNA | BT003862.1 | - | 1 - 2 | 298 | 71 | |
| BT005157.1 | - | 1 - 2 | 256 | 71 | ||
More Information
Set Membership: |
|

GRP16
Genomic Map