Assay Details
| Gene Symbol: PK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120899.3 | NP_196433.1 | 2 - 3 | 627 | 69 | |
| NM_180461.1 | NP_850792.1 | 3 - 4 | 424 | 69 | ||
| GenBank mRNA | AF083801.1 | - | 2 - 3 | 509 | 69 | |
| AF325057.2 | - | 2 - 3 | 532 | 69 | ||
| BT025237.1 | - | 2 - 3 | 364 | 69 | ||
| U97568.1 | - | 2 - 3 | 510 | 69 | ||
More Information
Set Membership: |
|

PK3
Genomic Map