Assay Details
| Gene Symbol: PK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120899.3 | NP_196433.1 | 3 - 4 | 764 | 116 | |
| NM_180461.1 | NP_850792.1 | 4 - 5 | 561 | 116 | ||
| GenBank mRNA | AF083801.1 | - | 3 - 4 | 646 | 116 | |
| AF325057.2 | - | 3 - 4 | 669 | 116 | ||
| BT025237.1 | - | 3 - 4 | 501 | 116 | ||
| U97568.1 | - | 3 - 4 | 647 | 116 | ||
More Information
Set Membership: |
|

PK3
Genomic Map