Assay Details
| Gene Symbol: NF-YA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343248.1 | NP_001318552.1 | 4 - 5 | 639 | 92 | |
| NM_121287.4 | NP_568282.1 | 4 - 5 | 529 | 92 | ||
| NM_180480.3 | NP_850811.1 | 3 - 4 | 704 | 92 | ||
| NM_203044.4 | NP_974773.1 | 3 - 4 | 774 | 92 | ||
| NM_203045.2 | NP_974774.1 | 4 - 5 | 611 | 92 | ||
| GenBank mRNA | AK317299.1 | - | 4 - 5 | 612 | 92 | |
| AX765878.1 | - | 2 - 3 | 377 | 92 | ||
| BT025801.1 | - | 2 - 3 | 380 | 92 | ||
| Y13720.1 | - | 656 | 92 | |||
More Information
Set Membership: |
|

NF-YA1
Genomic Map