Assay Details
| Gene Symbol: NF-YA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343248.1 | NP_001318552.1 | 5 - 6 | 705 | 66 | |
| NM_121287.4 | NP_568282.1 | 5 - 6 | 595 | 66 | ||
| NM_180480.3 | NP_850811.1 | 4 - 5 | 770 | 66 | ||
| NM_203044.4 | NP_974773.1 | 4 - 5 | 840 | 66 | ||
| NM_203045.2 | NP_974774.1 | 5 - 6 | 677 | 66 | ||
| GenBank mRNA | AK317299.1 | - | 5 - 6 | 678 | 66 | |
| AX765878.1 | - | 3 - 4 | 443 | 66 | ||
| BT025801.1 | - | 3 - 4 | 446 | 66 | ||
| Y13720.1 | - | 722 | 66 | |||
More Information
Set Membership: |
|

NF-YA1
Genomic Map