Assay Details
| Gene Symbol: NF-YA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343248.1 | NP_001318552.1 | 6 - 7 | 871 | 64 | |
| NM_121287.4 | NP_568282.1 | 6 - 7 | 761 | 64 | ||
| NM_180480.3 | NP_850811.1 | 5 - 6 | 936 | 64 | ||
| NM_203044.4 | NP_974773.1 | 5 - 6 | 1006 | 64 | ||
| NM_203045.2 | NP_974774.1 | 6 - 7 | 843 | 64 | ||
| GenBank mRNA | AK317299.1 | - | 6 - 7 | 844 | 64 | |
| AX765878.1 | - | 4 - 5 | 609 | 64 | ||
| BT025801.1 | - | 4 - 5 | 612 | 64 | ||
| Y13720.1 | - | 888 | 64 | |||
More Information
Set Membership: |
|

NF-YA1
Genomic Map