Assay Details
| Gene Symbol: PPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343294.1 | NP_001330304.1 | 3 - 4 | 949 | 74 | |
| NM_001343295.1 | NP_001330305.1 | 3 - 4 | 882 | 74 | ||
| NM_121383.4 | NP_196884.1 | 3 - 4 | 992 | 74 | ||
| NM_180484.3 | NP_850815.1 | 2 - 3 | 941 | 74 | ||
| GenBank mRNA | AK317089.1 | - | 2 - 3 | 696 | 74 | |
| AY056386.1 | - | 3 - 4 | 747 | 74 | ||
| AY070736.1 | - | 3 - 4 | 788 | 74 | ||
| AY149934.1 | - | 1 - 2 | 630 | 74 | ||
More Information
Set Membership: |
|

PPH
Genomic Map