Assay Details
| Gene Symbol: PPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343294.1 | NP_001330304.1 | 4 - 5 | 1170 | 90 | |
| NM_001343295.1 | NP_001330305.1 | 4 - 5 | 1103 | 90 | ||
| NM_121383.4 | NP_196884.1 | 4 - 5 | 1213 | 90 | ||
| NM_180484.3 | NP_850815.1 | 3 - 4 | 1162 | 90 | ||
| GenBank mRNA | AK317089.1 | - | 3 - 4 | 917 | 90 | |
| AY056386.1 | - | 4 - 5 | 968 | 90 | ||
| AY070736.1 | - | 4 - 5 | 1009 | 90 | ||
| AY149934.1 | - | 2 - 3 | 851 | 90 | ||
More Information
Set Membership: |
|

PPH
Genomic Map