Assay Details
| Gene Symbol: PPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343294.1 | NP_001330304.1 | 6 - 7 | 1517 | 55 | |
| NM_121383.4 | NP_196884.1 | 6 - 7 | 1560 | 55 | ||
| NM_180484.3 | NP_850815.1 | 5 - 6 | 1509 | 55 | ||
| GenBank mRNA | AK317089.1 | - | 5 - 6 | 1264 | 55 | |
| AY056386.1 | - | 6 - 7 | 1315 | 55 | ||
| AY070736.1 | - | 6 - 7 | 1356 | 55 | ||
| AY149934.1 | - | 4 - 5 | 1198 | 55 | ||
More Information
Set Membership: |
|

PPH
Genomic Map