Assay Details
| Gene Symbol: ARIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121938.3 | NP_197434.1 | 15 - 16 | 1690 | 155 | |
| NM_180521.3 | NP_850852.1 | 16 - 17 | 1876 | 155 | ||
More Information
Set Membership: |
|

ARIA
Genomic Map