Assay Details
| Gene Symbol: ARIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121938.3 | NP_197434.1 | 3 - 4 | 650 | 113 | |
| NM_180521.3 | NP_850852.1 | 3 - 4 | 689 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARIA| Gene Symbol: ARIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121938.3 | NP_197434.1 | 3 - 4 | 650 | 113 | |
| NM_180521.3 | NP_850852.1 | 3 - 4 | 689 | 113 | ||
Set Membership: |
|