Assay Details
| Gene Symbol: ARIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121938.3 | NP_197434.1 | 9 - 10 | 1249 | 136 | |
| NM_180521.3 | NP_850852.1 | 9 - 10 | 1288 | 136 | ||
More Information
Set Membership: |
|

ARIA
Genomic Map