Assay Details
| Gene Symbol: PYR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036865.1 | NP_001031942.1 | 5 - 6 | 558 | 78 | |
| NM_001343973.1 | NP_001332394.1 | 5 - 6 | 775 | 78 | ||
| NM_180536.3 | NP_850867.1 | 5 - 6 | 599 | 78 | ||
| NM_180537.3 | NP_850868.1 | 5 - 6 | 612 | 78 | ||
| GenBank mRNA | AF000147.1 | - | 5 - 6 | 480 | 78 | |
| BT029445.1 | - | 4 - 5 | 420 | 78 | ||
More Information
Set Membership: |
|

PYR6
Genomic Map