Assay Details
| Gene Symbol: PYR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343973.1 | NP_001332394.1 | 7 - 8 | 959 | 114 | |
| NM_180537.3 | NP_850868.1 | 7 - 8 | 796 | 114 | ||
| GenBank mRNA | AF000147.1 | - | 7 - 8 | 664 | 114 | |
More Information
Set Membership: |
|

PYR6
Genomic Map