Assay Details
Gene Symbol: AT5G55132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180595.2 | NP_850926.1 | 2 - 3 | 158 | 73 | |
GenBank mRNA | AK118018.1 | - | 2 - 3 | 158 | 73 | |
BT004662.1 | - | 1 - 2 | 67 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
