Assay Details
| Gene Symbol: CER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331327.1 | NP_001320548.1 | 7 - 8 | 1737 | 100 | |
| NM_001331328.1 | NP_001320547.1 | 6 - 7 | 2351 | 100 | ||
| NM_100101.4 | NP_171723.2 | 8 - 9 | 1938 | 100 | ||
| NM_180601.2 | NP_850932.2 | 7 - 8 | 1732 | 100 | ||
| GenBank mRNA | AK226895.1 | - | 7 - 8 | 1716 | 100 | |
More Information
Set Membership: |
|

CER1
Genomic Map