Assay Details
| Gene Symbol: SR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180603.1 | NP_850934.1 | 10 - 11 | 855 | 145 | |
| GenBank mRNA | AY128338.1 | - | 10 - 11 | 849 | 145 | |
More Information
Set Membership: |
|

SR34
Genomic Map