Assay Details
| Gene Symbol: MGP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331783.1 | NP_001320197.1 | 3 - 4 | 293 | 107 | |
| NM_100739.6 | NP_172342.3 | 5 - 6 | 657 | 107 | ||
| NM_180609.4 | NP_850940.1 | 5 - 6 | 657 | 107 | ||
| GenBank mRNA | AY064135.1 | - | 5 - 6 | 531 | 107 | |
| AY124807.1 | - | 5 - 6 | 445 | 107 | ||
| KJ139019.1 | - | 5 - 6 | 445 | 107 | ||
More Information
Set Membership: |
|

MGP2
Genomic Map