Assay Details
| Gene Symbol: SCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198248.1 | NP_001185177.1 | 24 - 25 | 2855 | 126 | |
| NM_001333364.1 | NP_001320175.1 | 23 - 24 | 3444 | 126 | ||
| NM_180628.4 | NP_850959.1 | 24 - 25 | 2893 | 126 | ||
| GenBank mRNA | AY082605.1 | - | 24 - 25 | 2700 | 126 | |
More Information
Set Membership: |
|

SCD1
Genomic Map