Assay Details
| Gene Symbol: SCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198248.1 | NP_001185177.1 | 8 - 9 | 1265 | 97 | |
| NM_001333364.1 | NP_001320175.1 | 8 - 9 | 1280 | 97 | ||
| NM_103797.4 | NP_175333.3 | 8 - 9 | 1217 | 97 | ||
| NM_180628.4 | NP_850959.1 | 8 - 9 | 1303 | 97 | ||
| GenBank mRNA | AY082605.1 | - | 8 - 9 | 1110 | 97 | |
More Information
Set Membership: |
|

SCD1
Genomic Map