Assay Details
| Gene Symbol: PREP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333396.1 | NP_001320455.1 | 13 - 14 | 2365 | 69 | |
| NM_001333397.1 | NP_001320454.1 | 17 - 18 | 3046 | 69 | ||
| NM_103851.3 | NP_175386.2 | 17 - 18 | 3075 | 69 | ||
| NM_180630.2 | NP_850961.1 | 17 - 18 | 3188 | 69 | ||
| NM_180631.2 | NP_850962.1 | 17 - 18 | 3188 | 69 | ||
| GenBank mRNA | AY074305.1 | - | 17 - 18 | 3075 | 69 | |
| BT004376.1 | - | 17 - 18 | 3030 | 69 | ||
More Information
Set Membership: |
|

PREP2
Genomic Map