Assay Details
| Gene Symbol: PREP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333397.1 | NP_001320454.1 | 1 - 2 | 333 | 135 | |
| NM_103851.3 | NP_175386.2 | 1 - 2 | 333 | 135 | ||
| NM_180630.2 | NP_850961.1 | 1 - 2 | 446 | 135 | ||
| NM_180631.2 | NP_850962.1 | 1 - 2 | 446 | 135 | ||
| GenBank mRNA | AY074305.1 | - | 1 - 2 | 333 | 135 | |
| BT004376.1 | - | 1 - 2 | 288 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001333397.1 transcripts.
Set Membership: |
|

PREP2
Genomic Map