Assay Details
| Gene Symbol: BGLU46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180637.2 | NP_850968.1 | 10 - 11 | 1623 | 144 | |
| NM_202338.2 | NP_974067.1 | 5 - 6 | 1212 | 144 | ||
More Information
Set Membership: |
|

BGLU46
Genomic Map