Assay Details
| Gene Symbol: BGLU46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180637.2 | NP_850968.1 | 4 - 5 | 725 | 170 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLU46| Gene Symbol: BGLU46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180637.2 | NP_850968.1 | 4 - 5 | 725 | 170 | |
Set Membership: |
|