Assay Details
| Gene Symbol: AT5G04120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120494.4 | NP_196032.1 | 2 - 3 | 293 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G04120| Gene Symbol: AT5G04120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120494.4 | NP_196032.1 | 2 - 3 | 293 | 65 | |
Set Membership: |
|