Assay Details
| Gene Symbol: NAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120523.4 | NP_196061.1 | 1 - 2 | 459 | 129 | |
| GenBank mRNA | AY099609.1 | - | 1 - 2 | 332 | 129 | |
| AY128859.1 | - | 1 - 2 | 163 | 129 | ||
More Information
Set Membership: |
|

NAC2
Genomic Map