Assay Details
| Gene Symbol: CAT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342790.1 | NP_001330125.1 | 2 - 3 | 1302 | 69 | |
| NM_001342791.1 | NP_001330124.1 | 1 - 2 | 1252 | 69 | ||
| NM_120559.4 | NP_196097.1 | 2 - 3 | 1469 | 69 | ||
| GenBank mRNA | AF360186.1 | - | 2 - 3 | 1010 | 69 | |
| AY040001.1 | - | 2 - 3 | 972 | 69 | ||
| AY085578.1 | - | 2 - 3 | 1010 | 69 | ||
More Information
Set Membership: |
|

CAT6
Genomic Map