Assay Details
Gene Symbol: CAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342791.1 | NP_001330124.1 | 2 - 3 | 1485 | 74 | |
NM_120559.4 | NP_196097.1 | 3 - 4 | 1702 | 74 | ||
GenBank mRNA | AF360186.1 | - | 3 - 4 | 1243 | 74 | |
AY040001.1 | - | 3 - 4 | 1205 | 74 | ||
AY085578.1 | - | 3 - 4 | 1243 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
