Assay Details
| Gene Symbol: EMB2771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342824.1 | NP_001318485.1 | 10 - 11 | 1890 | 76 | |
| NM_001342825.1 | NP_001331419.1 | 11 - 12 | 3048 | 76 | ||
| NM_001342826.1 | NP_001331420.1 | 9 - 10 | 1552 | 76 | ||
More Information
Set Membership: |
|

EMB2771
Genomic Map