Assay Details
| Gene Symbol: EMB2771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342824.1 | NP_001318485.1 | 16 - 17 | 3023 | 87 | |
| NM_001342825.1 | NP_001331419.1 | 17 - 18 | 4181 | 87 | ||
| NM_001342826.1 | NP_001331420.1 | 15 - 16 | 2685 | 87 | ||
More Information
Set Membership: |
|

EMB2771
Genomic Map