Assay Details
| Gene Symbol: EMB2771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342824.1 | NP_001318485.1 | 22 - 23 | 3551 | 75 | |
| NM_001342825.1 | NP_001331419.1 | 23 - 24 | 4709 | 75 | ||
| NM_001342826.1 | NP_001331420.1 | 21 - 22 | 3213 | 75 | ||
More Information
Set Membership: |
|

EMB2771
Genomic Map