Assay Details
| Gene Symbol: EMB2771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342824.1 | NP_001318485.1 | 7 - 8 | 1186 | 163 | |
| NM_001342825.1 | NP_001331419.1 | 8 - 9 | 2344 | 163 | ||
| NM_001342826.1 | NP_001331420.1 | 6 - 7 | 848 | 163 | ||
More Information
Set Membership: |
|

EMB2771
Genomic Map