Assay Details
| Gene Symbol: NFXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120648.6 | NP_196185.4 | 7 - 8 | 1835 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFXL2| Gene Symbol: NFXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120648.6 | NP_196185.4 | 7 - 8 | 1835 | 142 | |
Set Membership: |
|