Assay Details
| Gene Symbol: ASA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203302.1 | NP_001190231.1 | 2 - 3 | 473 | 63 | |
| NM_120655.4 | NP_196192.1 | 2 - 3 | 496 | 63 | ||
More Information
Set Membership: |
|

ASA1
Genomic Map