Assay Details
| Gene Symbol: ASA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203302.1 | NP_001190231.1 | 6 - 7 | 1321 | 87 | |
| NM_120655.4 | NP_196192.1 | 7 - 8 | 1257 | 87 | ||
More Information
Set Membership: |
|

ASA1
Genomic Map