Assay Details
| Gene Symbol: PIRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120667.5 | NP_196204.1 | 2 - 3 | 1052 | 129 | |
| GenBank mRNA | AK228975.1 | - | 2 - 3 | 956 | 129 | |
| AY849571.1 | - | 2 - 3 | 887 | 129 | ||
| BT015883.1 | - | 2 - 3 | 892 | 129 | ||
| BT020196.1 | - | 2 - 3 | 887 | 129 | ||
More Information
Set Membership: |
|

PIRL1
Genomic Map