Assay Details
| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342841.1 | NP_001318487.1 | 3 - 4 | 868 | 136 | |
| NM_001342842.1 | NP_001331576.1 | 3 - 4 | 697 | 136 | ||
| NM_001342843.1 | NP_001331580.1 | 2 - 3 | 803 | 136 | ||
| NM_001342844.1 | NP_001331579.1 | 2 - 3 | 985 | 136 | ||
| NM_001342845.1 | NP_001331578.1 | 1 - 2 | 204 | 136 | ||
| NM_001342846.1 | NP_001331577.1 | 3 - 4 | 658 | 136 | ||
| NM_120675.5 | NP_568159.1 | 3 - 4 | 697 | 136 | ||
| GenBank mRNA | AK229272.1 | - | 3 - 4 | 521 | 136 | |
| AY118140.1 | - | 2 - 3 | 301 | 136 | ||
| BT024499.1 | - | 2 - 3 | 301 | 136 | ||
More Information
Set Membership: |
|

GC1
Genomic Map