Assay Details
| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342841.1 | NP_001318487.1 | 5 - 6 | 1046 | 97 | |
| NM_001342842.1 | NP_001331576.1 | 5 - 6 | 875 | 97 | ||
| NM_001342843.1 | NP_001331580.1 | 4 - 5 | 981 | 97 | ||
| NM_001342844.1 | NP_001331579.1 | 4 - 5 | 1163 | 97 | ||
| NM_001342845.1 | NP_001331578.1 | 3 - 4 | 382 | 97 | ||
| NM_001342846.1 | NP_001331577.1 | 5 - 6 | 836 | 97 | ||
| NM_120675.5 | NP_568159.1 | 5 - 6 | 875 | 97 | ||
| GenBank mRNA | AK229272.1 | - | 5 - 6 | 699 | 97 | |
| AY118140.1 | - | 4 - 5 | 479 | 97 | ||
| BT024499.1 | - | 4 - 5 | 479 | 97 | ||
More Information
Set Membership: |
|

GC1
Genomic Map