Assay Details
| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342841.1 | NP_001318487.1 | 9 - 10 | 1349 | 140 | |
| NM_001342842.1 | NP_001331576.1 | 8 - 9 | 1423 | 140 | ||
| NM_001342843.1 | NP_001331580.1 | 8 - 9 | 1284 | 140 | ||
| NM_001342844.1 | NP_001331579.1 | 8 - 9 | 1466 | 140 | ||
| NM_001342845.1 | NP_001331578.1 | 7 - 8 | 685 | 140 | ||
| NM_120675.5 | NP_568159.1 | 9 - 10 | 1178 | 140 | ||
| GenBank mRNA | AK229272.1 | - | 9 - 10 | 1002 | 140 | |
More Information
Set Membership: |
|

GC1
Genomic Map