Assay Details
| Gene Symbol: ROPGEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342847.1 | NP_001332731.1 | 1 - 2 | 484 | 105 | |
| NM_120676.3 | NP_196213.2 | 1 - 2 | 484 | 105 | ||
More Information
Set Membership: |
|

ROPGEF5
Genomic Map