Assay Details
| Gene Symbol: ROPGEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120676.3 | NP_196213.2 | 6 - 7 | 1490 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROPGEF5| Gene Symbol: ROPGEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120676.3 | NP_196213.2 | 6 - 7 | 1490 | 78 | |
Set Membership: |
|