Assay Details
| Gene Symbol: DFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036763.2 | NP_001031840.1 | 14 - 15 | 1646 | 71 | |
| NM_120680.3 | NP_196217.2 | 15 - 16 | 1588 | 71 | ||
More Information
Set Membership: |
|

DFB
Genomic Map