Assay Details
| Gene Symbol: DFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036763.2 | NP_001031840.1 | 1 - 2 | 296 | 186 | |
| NM_120680.3 | NP_196217.2 | 2 - 3 | 238 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001036763.2 transcripts.
Set Membership: |
|

DFB
Genomic Map