Assay Details
| Gene Symbol: DFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036763.2 | NP_001031840.1 | 3 - 4 | 476 | 126 | |
| NM_120680.3 | NP_196217.2 | 4 - 5 | 418 | 126 | ||
More Information
Set Membership: |
|

DFB
Genomic Map