Assay Details
| Gene Symbol: DFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036763.2 | NP_001031840.1 | 6 - 7 | 731 | 83 | |
| NM_120680.3 | NP_196217.2 | 7 - 8 | 673 | 83 | ||
More Information
Set Membership: |
|

DFB
Genomic Map